Learn More
Medchemexpress LLC Darunavir Ethanolate | 635728-49-3 | 99.9% | 593.73 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY17041200MG
Encompass_Preferred
Darunavir ethanolate (TMC114 Ethanolate) is a potent HIV protease inhibitor used to treat and prevent HIV/AIDS. It has a Ki of 1 nM for wild type HIV-1 protease.
  • Broad-spectrum potent inhibitor active against HIV-1 clinical isolates with minimal cytotoxicity.
  • Forms hydrogen bonds with the conserved main-chain atoms of Asp29 and Asp30 of the protease, critical for potency against HIV isolates resistant to multiple protease inhibitors.
  • Shows greater potency than other inhibitors in MT-2 cells.
  • Primarily metabolized by hepatic cytochrome P450 (CYP) enzymes, mainly CYP3A.
  • Is effective against wild-type and PI-resistant HIV.
  • Has an oral bioavailability of 37%, which increases to 82% when combined with ritonavir.
Catalog No. 50-003-85874