Learn More
Medchemexpress LLC Dasatinib | 302962-49-8 | MFCD11046566 | ≥98.0% | 488.01 | C22H26ClN7O2S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1018110MM 1ML
Encompass_Preferred
Dasatinib is a potent, ATP-competitive small-molecule inhibitor of Bcr-Abl and Src family kinases used in research to probe kinase signaling, assess antitumor activity, and as a reference compound in biochemical and cell-based studies. It is typically handled as a solid for preparation of concentrated stock solutions or provided by suppliers as prepared solutions for experimental use.
  • Potent dual Src/Bcr-Abl inhibition (IC50 <1.0 nM for Bcr-Abl, ~0.5 nM for Src).
  • Suitable for biochemical assays and cell-based pathway studies.
  • Includes guidance for preparing 1-10 mM stock solutions from solid material.
  • Molecular weight 488.01; formula C22H26ClN7O2S; CAS 302962-49-8.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-48860