Learn More
Selleck Chemical LLC Dasatinib 100mg 302962-49-8 BMS-354825
SDP
Supplier:  Selleck Chemical LLC S1021100MG
Encompass_Preferred
Dasatinib (BMS-354825) is a novel, potent and multi-targeted inhibitor that targets Abl, Src and c-Kit, with IC50 of <1 nM, 0.8 nM and 79 nM in cell-free assays, respectively. Dasatinib induces autophagy and apoptosis with anti-tumor activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-743-0