Learn More
Medchemexpress LLC Dasatinib carbaldehyde | 2112837-79-1 | 98.5% | 471.96 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11185750MG
Encompass_Preferred
Dasatinib carbaldehyde (BMS-354825 carbaldehyde) is a Dasatinib (ABL inhibitor) based moiety that binds to an IAP ligand via a linker to form SNIPER. It is intended for research use only.
  • Binds to IAP ligand via a linker
  • Forms SNIPER
  • Solid appearance
  • White to off-white color
  • Soluble in DMSO (33.33 mg/mL)
  • Stable for 3 years at -20°C as powder
  • Stable for 6 months at -80°C in solvent
Catalog No. 50-004-38906