Learn More
Medchemexpress LLC Dasatinib carboxylic acid | 910297-53-9 | MFCD18252608 | 99.6% | C22H24ClN7O3S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13166910MG
Encompass_Preferred
Dasatinib metabolite M6 (Dasatinib carboxylic acid) is an oxidative metabolite of the tyrosine kinase inhibitor dasatinib supplied as a research-grade reference standard for in vitro assays, metabolic profiling, and analytical method development.
  • Provided as a high-purity analytical standard (99.6%).
  • Available in multiple solid quantities and as a 10 mM solution in DMSO.
  • Suitable for metabolic identification and LC-MS method development.
  • Molecular formula C22H24ClN7O3S; molecular weight 501.99 g·mol⁻1.
Catalog No. 50-002-59161