Learn More
Medchemexpress LLC Dasatinib-d4 | 1160297-08-4 | 97.5% | 492.03 g/mol | C22H22D4ClN7O2S | 1 MG

Supplier: Medchemexpress LLC HY10181S11MG
Dasatinib-d4 is a deuterium-labeled analogue of the tyrosine kinase inhibitor dasatinib, provided as a high-purity analytical reference for research use. It is formulated for use as an internal standard in quantitative LC-MS/MS assays and method development workflows.
- Deuterium-labeled internal standard for quantitative analysis
- High purity suitable for analytical applications
- Available in small reference pack sizes for trace-level studies
- Molecular weight and formula provided for accurate characterization
- CAS provided for unambiguous compound identification
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.