Learn More
Medchemexpress LLC Dasatinib intermediate-1 | 302964-08-5 | ≥98.0% | 394.28 | 25 G
Supplier: Medchemexpress LLC 25GHY41940
Dasatinib intermediate-1 is an intermediate used in the synthesis of Dasatinib. Dasatinib itself is a highly potent, ATP competitive, orally active dual Src/Bcr-Abl inhibitor with significant antitumor activity.
- Intermediate in the synthesis of Dasatinib.
- It is a highly potent, ATP competitive, orally active dual Src/Bcr-Abl inhibitor with potent antitumor activity.