Learn More
Medchemexpress LLC Dasatinib intermediate-1 | 302964-08-5 | ≥98.0% | 394.28 | 50 G
SDP
Supplier:  Medchemexpress LLC HY4194050G
Encompass_Preferred
Dasatinib intermediate-1 is a compound used in the synthesis of Dasatinib, a highly potent, ATP competitive, orally active dual Src/Bcr-Abl inhibitor that exhibits potent antitumor activity. It is intended for research use only and not for patient use.
  • Used in the synthesis of Dasatinib
  • Off-white to light yellow solid
  • For research use only
  • Not for patient use
Catalog No. 50-004-03441