Learn More
CPC Scientific H-His-Ser-Gln-Gly-Thr-Phe-Thr-Ser-Asp-Tyr-Ser-Lys-Tyr-Leu-Asp-Aib-Ala-Arg-Ala-Glu-Glu-Phe-Val-Lys-Trp-Leu-Glu-Ser-Thr-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific GLUC020B
Encompass
Metabolically stable analog of glucagonONE-LETTER SEQUENCE: HSQGTFTSDYSKYLDXARAEEFVKWLESTMOLECULAR FORMULA: C152H222N38O50MOLECULAR WEIGHT:3381.66STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1544300-84-6],
Catalog No. 50-210-9391