Learn More
Medchemexpress LLC Dasminapant | 1570231-89-8 | 99.7% | 1157.40 | C60H72N10O10S2 | 1MG
Supplier: Medchemexpress LLC HY1255931MG
Dasminapant (APG-1387) is a bivalent SMAC mimetic and inhibitor of apoptosis protein (IAP) antagonist provided for research use. It promotes degradation of cIAP-1 and XIAP, activates caspase-3, and induces apoptosis in preclinical cancer models, supporting studies in hepatocellular, ovarian, and nasopharyngeal carcinoma.
- Bivalent SMAC mimetic that antagonizes IAP family proteins.
- Promotes degradation of cIAP-1 and XIAP and triggers caspase activation.
- Used in preclinical studies of various solid tumors.
- High reported purity suitable for research applications.
- Available in small milligram pack sizes for experimental use.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.