Learn More
TARGETMOL CHEMICALS INC DASOTRALINE HYDROCHLORID 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T352650MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dasotraline hydrochloride (SEP-225289 hydrochloride) (SEP-225289 hydrochloride) is a triple reuptake inhibitor that inhibits dopamine norepinephrine and serotonin transporters with IC50 values of 4 6 and 11 nM respectively. purity: 99%
Catalog No. 50-258-0228