Learn More
Medchemexpress LLC Dasotraline hydrochloride | 675126-08-6 | >98.0% | 328.66 g/mol | C16H16Cl3N | 5 MG
SDP
Supplier:  Medchemexpress LLC HY12850A5MG
Encompass_Preferred
Dasotraline hydrochloride is the hydrochloride salt of dasotraline (SEP-225289), a triple reuptake inhibitor that blocks dopamine, norepinephrine, and serotonin transporters. It is provided for research use in biochemical and pharmacological studies of monoamine transporter inhibition.
  • Triple reuptake inhibitor activity at DAT, NET, and SERT with reported low-nanomolar IC50 values.
  • High purity suitable for research and analytical applications.
  • Available in small mass quantities for preparation of stock solutions.
  • Analytical data (HPLC, NMR) provided in accompanying documentation.
  • Suitable as a reference compound in transporter and pharmacokinetic assays.
Catalog No. 50-002-64029