Learn More
Medchemexpress LLC Dasotraline hydrochloride | 675126-08-6 | 99.0% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12850A10MG
Encompass_Preferred
Dasotraline hydrochloride is a triple reuptake inhibitor that effectively blocks dopamine, norepinephrine, and serotonin transporters. This compound is intended for research use only.
  • Blocks dopamine, norepinephrine, and serotonin transporters
  • Exhibits IC50 values of 4 nM (dopamine), 6 nM (norepinephrine), and 11 nM (serotonin)
Catalog No. 50-003-41838