Learn More
Cole-Parmer Traceable Memory-Loc™ Calibrated Datalogging Thermometer; 1 Vaccine Bottle Probe
Supplier:  Cole-Parmer 9876765
Encompass_Preferred
Raw data is locked and may not be cleared or changed on base unitStore up to 10 unique alarm events, memory can be cleared after downloading dataSmart-Alarm™ with visual/audible alarm continues to alert even if the unit returns to nonalarm conditionsTriple-display—shows high, low, and current temperature simultaneouslyIncludes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
Catalog No. NC3054786