Learn More
Medchemexpress LLC Datelliptium chloride | 105118-14-7 | 397.94 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYU0033750MG
Encompass_Preferred
Datelliptium chloride is a DNA-intercalating agent derived from ellipticine, with anti-tumor activities.
  • DNA-intercalating agent
  • Derived from ellipticine
  • Exhibits anti-tumor activities
  • Shows significantly cytotoxic effects in suspension and primary cultures of rat hepatocytes (100 μM after 2 hr treatment)
  • Mobilizes glycogen in rat systems in vitro concentration-dependently
  • Leads to a concentration-dependent decrease in gluconeogenesis from lactate in cells
  • Datelliptium acetate (a related form) is selectively cytotoxic for solid tumors over leukemia L1210
  • Datelliptium acetate shows potent inhibitory activities in vivo against various murine solid tumors upon intravenous administration
Catalog No. 50-004-47139