Learn More
Medchemexpress LLC Datpt | 2891582-75-3 | 99.5% | 495.06 g·mol⁻¹ | C24H39ClN6O3 | 5 MG

Supplier: Medchemexpress LLC HY1453075MG
DATPT is a peptide-mimetic small molecule that blocks the SNX9-p47phox interaction, suppressing reactive oxygen species and inflammatory cytokine production. It exhibits reported anti-inflammatory and antibacterial activity and is supplied for preclinical research and formulation studies.
- High purity: 99.5%.
- Molecular formula: C24H39ClN6O3; molecular weight: 495.06 g·mol⁻¹.
- Store solid at 4°C, sealed and away from moisture; in solvent store at -80°C (6 months) or -20°C (1 month).
- Soluble in DMSO at 100 mg/mL; in vivo formulations achieve ~2.5 mg/mL with provided protocols.
- Intended for preclinical research, including inflammatory and sepsis studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.