Learn More
Medchemexpress LLC Daunomycinone 10mg | 21794-55-8 | 10 MG

Supplier: Medchemexpress LLC HY11465810MG
Daunomycinone is an anthracycline metabolite used as a research chemical and intermediate in anticancer studies. It is supplied as a high-purity solid and is soluble in dimethyl sulfoxide (DMSO) for in vitro applications.
- High purity: 98.9%.
- Molecular weight 398.36.
- Chemical formula C21H18O8.
- Physical state: solid, orange to red.
- Soluble in DMSO at 100 mg/mL; ultrasonic assistance may be required.
- Provided in a 10 mg package suitable for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.