Learn More
Medchemexpress LLC Daunomycinone 50mg | 21794-55-8 | 398.36 g/mol | C21H18O8 | 50 MG

Supplier: Medchemexpress LLC HY11465850MG
Daunomycinone (NSC-109351) is an analytical standard and metabolite of daunomycin used as a reference material for research and analytical applications, including leukemia studies. The compound is supplied as a characterized solid with documented purity and accompanying quality and safety documentation.
- Analytical standard for daunomycin metabolite studies.
- Intended for research and analytical applications.
- CAS number 21794-55-8.
- Molecular formula C21H18O8.
- Molecular weight 398.36 g/mol.
- Purity 98.9% (as listed).
- Supplied as a solid; available in a 50 MG package.
- Certificate of Analysis and safety data sheet available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.