Learn More
Medchemexpress LLC Daunorubicin-13C,d3 | 20830-81-3 | MFCD00866340 | 98.0% | 531.53 | C2613CH26D3NO10 | 250 UG
SDP
Supplier:  Medchemexpress LLC HYW654130250UG
Encompass_Preferred
Isotopically labeled daunorubicin (13C and d3) supplied as a low-quantity analytical standard for research use. It is intended for use as an internal standard and tracer in mass spectrometry, pharmacokinetic, metabolic, and mechanistic studies involving topoisomerase II inhibitors.
  • Isotopically labeled with 13C and d3 for unambiguous mass-spectrometry detection.
  • Suitable as an internal standard for LC-MS quantitation and method development.
  • Appropriate for pharmacokinetic and metabolic tracing studies.
  • Provided in a small 250 μg amount to support high-sensitivity assays.
  • Documentation available; certificate of analysis provides purity and handling information.
Catalog No. 50-002-84242