Learn More
Cayman Chemical DaunorubIcIn-13C-d3 250ug
SDP
Supplier:  Cayman Chemical 29101250 UG
Encompass_Preferred
An internal standard for the quantification of daunorubicin by GC- or LC-MS
Catalog No. 50-294-0306