Learn More
Selleck Chemical LLC Daunorubicin E8164-5MG
SDP
Supplier:  Selleck Chemical LLC E81645MG
Encompass_Preferred
Daunorubicin (Daunomycin) is a topoisomerase II inhibitor with potent anti-tumor activity Daunorubicin inhibits DNA and RNA synthesis Daunorubicin is a cytotoxin that inhibits cancer cell viability and induces apoptosis and necrosis Daunorubicin is also an anthracycline antibiotic Daunorubicin can be used in the research of infection and variety of cancers including leukemia non-Hodgkin lymphomas Ewing s sarcoma Wilms tumor
Catalog No. 50-000-41945