Learn More
Enzo Life Sciences Daunorubicin . hydrochloride (10mg). CAS: 23541-50-6
SDP
Supplier:  Enzo Life Sciences ALX380043M010
Encompass_Preferred
Anthracycline anticancer antibiotic. Induces DNA damage by intercalating into DNA and inhibiting topoisomerase I and II. Inhibits RNA and DNA synthesis. Induces DNA single strand breaks and apoptosis in tumor cells. Alternative name: Daunomycin . HCl. Purity: ≥90% (Assay). Appearance: Orange to red crystalline powder. Solubility: Soluble in water or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-8930