Learn More
Medchemexpress LLC Daunorubicin (hydrochloride) | 23541-50-6 | 99.7% | 25 MG

Supplier: Medchemexpress LLC 25MGHY13062
Daunorubicin hydrochloride is a potent topoisomerase II inhibitor with significant anti-tumor activity. This anthracycline antibiotic inhibits both DNA and RNA synthesis, functioning as a cytotoxin that reduces cancer cell viability and induces apoptosis and necrosis. It is utilized in research for various infections and cancers, including leukemia, non-Hodgkin lymphomas, Ewing's sarcoma, and Wilms' tumor. The compound is a pink to red solid, stable under recommended storage conditions, with a melting point of 190°C.
- Target: topoisomerase, DNA/RNA synthesis, ADC payload, bacterial, autophagy, apoptosis, antibiotic
- Research areas: infection, neurological disease, cancer
- CAS number: 23541-50-6
- Molecular formula: C27H30ClNO10
- Molecular weight: 563.98
- Appearance: pink to red solid
- Purity (LCMS): 99.7%
- Melting point: 190°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.