Learn More
TARGETMOL CHEMICALS INC DAPANSUTRILE 25MG
Supplier: TARGETMOL CHEMICALS INC T1505225MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dapansutrile is a potent selective and orally active NLRP3 inflammasome inhibitor with Anti-inflammatory and analgesic activity. purity: 98%