Learn More
Medchemexpress LLC Daurab2 | 3115765-38-0 | 1049.02 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY171769
Encompass_Preferred
dAurAB2 is a dual-degrading PROTAC that potently degrades Aurora-A and Aurora-B, reducing N-Myc levels in MYCN-amplified IMR32 neuroblastoma cells. It is suitable for the study of neuroblastoma.
  • Potently degrades Aurora-A and Aurora-B with DC50s of 59 nM and 39 nM, respectively.
  • Reduces N-Myc levels in MYCN-amplified IMR32 neuroblastoma cells.
  • Can be used for neuroblastoma studies.
  • A dual-degrading PROTAC.
Catalog No. 50-003-84406