Learn More
Medchemexpress LLC dAurAB2 | 3115765-38-0 | 1049.02 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1717695MG
Encompass_Preferred
dAurAB2 is a dual-degrading PROTAC that potently degrades Aurora-A and Aurora-B with DC50s of 59 nM and 39 nM, respectively. It reduces N-Myc levels in MYCN-amplified IMR32 neuroblastoma cells and can be used for the study of neuroblastoma.
  • Potently degrades Aurora-A and Aurora-B
  • Reduces N-Myc levels in MYCN-amplified IMR32 neuroblastoma cells
  • Can be used for the study of neuroblastoma
  • Product has not been fully validated for medical applications. For research use only.
Catalog No. 50-003-86513