Learn More
Agilent Technologies J&W DB-624 GC Column, 60 m, 0.53 mm, 3.00 µm, 7 inch cage
Supplier:  Agilent Technologies 1251364
Encompass
This is the most common GC column format compatible with Agilent 5890, 6890, 7820, 7890, 8860, and 8890 series GC systems and all non-Agilent GC systems with a similarly sized oven.
Catalog No. NC3626768