Learn More
Medchemexpress LLC DB2115 tertahydrochloride | 1366126-19-3 | 99.1% | 704.48 | C32H34Cl4N8O2 | 100MG
SDP
Supplier:  Medchemexpress LLC HY124676A100MG
Encompass_Preferred
DB2115 tertahydrochloride is a research-grade small-molecule inhibitor of the myeloid transcription factor PU.1. Supplied as the tertahydrochloride salt, it has been reported to inhibit PU.1-DNA binding, reduce tumor cell proliferation, and induce apoptosis in leukemia models.
  • Potent PU.1 inhibitor (IC50 ≈ 2.3 nM).
  • Provided as the tertahydrochloride salt for improved handling.
  • High reported purity (≈99.1%).
  • Molecular formula C32H34Cl4N8O2 and molecular weight 704.48 g/mol.
  • Supplied in small research-scale quantities (typical 100 mg).
  • Intended for research use only; not for human use.
Catalog No. 50-002-38332