Learn More
Medchemexpress LLC Db2115 tertahydrochloride | 1366126-19-3 | 99.1% | 704.48 g/mol | C32H34Cl4N8O2 | 25MG
SDP
Supplier:  Medchemexpress LLC HY124676A25MG
Encompass_Preferred
DB2115 tertahydrochloride is a small-molecule inhibitor of the myeloid transcription factor PU.1, supplied as the hydrochloride salt for research use. It is a blue to purple solid with molecular formula C32H34Cl4N8O2 and molecular weight 704.48 g/mol. The compound is provided in milligram pack sizes and is reported at high purity for biochemical and cellular assays.
  • Potent PU.1 inhibitor suitable for preclinical research.
  • High reported purity (≈99.1%).
  • Blue to purple solid; suitable for low-temperature storage.
  • Soluble in DMSO for assay preparation.
  • Supplied in small milligram pack sizes for screening and discovery work.
Catalog No. 50-002-36257