Learn More
Medchemexpress LLC DB772 50mg | 1451058-50-6 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11462150MG
Encompass_Preferred
DB772 hydrate is a research-use small-molecule inhibitor reported to inhibit bovine viral diarrhoea virus (BVDV) and to exhibit anti-prion activity. It is supplied as a hydrate solid for laboratory research and is not intended for clinical or veterinary use.
  • Hydrate solid; molecular formula C20H19ClN4O2.
  • Molecular weight 502.54 g·mol⁻1.
  • Reported activity: BVDV inhibitor and anti-prion effects.
  • Intended for laboratory research use only.
  • Available in small-scale quantities, including 50 mg packs.
Catalog No. 50-002-28501