Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dbco-acid | 1353016-70-2 | MFCD28064336 | 99.7% | 305.33 g/mol | C19H15NO3 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY42972100MG

Encompass_Preferred

Dbco-acid is a dibenzocyclooctyne carboxylic acid used as a click chemistry reagent for bioorthogonal conjugation and linker synthesis. It is supplied as a purified solid for research applications and for preparing DBCO-derived NHS esters and PEG conjugates.

  • Used for bioorthogonal click chemistry and linker synthesis.
  • High purity: 99.71%.
  • Molecular weight: 305.33 g/mol.
  • Chemical formula: C19H15NO3.
  • Available as 100 MG solid with accompanying datasheet and SDS.

Catalog No. 50-002-02037


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.