Learn More
Medchemexpress LLC Dbco-acid | 1353016-70-2 | MFCD28064336 | 99.7% | 305.33 g/mol | C19H15NO3 | 50 MG

Supplier: Medchemexpress LLC HY4297250MG
DBCO-acid is a dibenzocyclooctyne-based click chemistry reagent used for strain-promoted alkyne-azide cycloaddition (SPAAC) and for synthesis of linker and bioconjugation reagents. It is provided in multiple pack sizes and is typically supplied with supporting analytical data and documentation.
- Used for strain-promoted alkyne-azide cycloaddition (SPAAC)
- Acid functional group enables conjugation via standard coupling chemistry
- High purity suitable for synthesis and bioconjugation applications
- Molecular weight approximately 305.33 g/mol
- CAS number 1353016-70-2 for unambiguous identification
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.