Learn More
Medchemexpress LLC DBCO-amine | 1255942-06-3 | 99.1% | 276.34 | C18H16N2O | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW000423250MG
Encompass_Preferred
DBCO-amine is a DBCO-containing click chemistry reagent used for strain-promoted alkyne-azide cycloaddition (SPAAC) and bioconjugation, including antibody-drug conjugate linker synthesis. It is supplied at high reported purity and in multiple pack sizes for research and synthetic applications.
  • Used for strain-promoted alkyne-azide cycloaddition (SPAAC) and bioconjugation.
  • Suitable for antibody-drug conjugate linker synthesis.
  • Reported purity 99.1%.
  • Molecular formula C18H16N2O; molecular weight 276.34.
  • Available in multiple pack sizes including 250 MG.
Catalog No. 50-002-02740