Learn More
Apexbio Technology LLC DBCO-C6-acid 1425485-72-8 250mg
SDP
Supplier:  Apexbio Technology LLC BA2469250
Encompass_Preferred
DBCO-C6-acid (CAS 1425485-72-8) is a non-cleavable linker featuring a dibenzocyclooctyne (DBCO) moiety primarily utilized in bioconjugation strategies such as antibody drug conjugate (ADC) construction This molecule engages in strain-promoted azide alkyne cycloaddition (SPAAC) enabling efficient copper-free click reactions with azide-containing biomolecules DBCO-C6-acid facilitates the site-specific attachment of payloads to antibodies or proteins without requiring metal catalysts thus preserving biological activity It is also used in the synthesis of carmaphycin analogs and other bioactive compounds for research applications involving targeted delivery and molecular labeling
Catalog No. 50-000-89383