Learn More
Medchemexpress LLC DBCO-C6-acid | 1425485-72-8 | MFCD22380737 | 98.9% | 333.38 | C21H19NO3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY121805100MG
Encompass_Preferred
DBCO-C6-acid is a dibenzocyclooctyne (DBCO)-based click chemistry reagent featuring a six-carbon spacer arm. It is supplied as an off-white to light brown solid with high purity for use in bioconjugation and antibody-drug conjugate linker synthesis.
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) click reactions.
  • Six-carbon spacer arm improves derivatization efficiency.
  • High purity (98.9%) suitable for research and synthesis workflows.
  • Soluble in DMSO at high concentration; ultrasonic assistance recommended.
  • Available in multiple laboratory-friendly sizes for scale flexibility.
Catalog No. 50-002-02193