Learn More
Medchemexpress LLC DBCO-C6-acid | 1425485-72-8 | MFCD22380737 | 98.9% | 333.38 | C21H19NO3 | 250 MG
SDP
Supplier:  Medchemexpress LLC HY121805250MG
Encompass_Preferred
DBCO-C6-acid is a DBCO-based click-chemistry reagent and non-cleavable antibody-drug conjugate (ADC) linker designed for strain-promoted alkyne-azide cycloaddition (SPAAC). It facilitates copper-free bioconjugation to azide-containing molecules and is used in the synthesis of linker-payload constructs such as carmaphycin analogues.
  • Enables copper-free SPAAC click reactions with azide-functionalized partners.
  • Contains a six-carbon spacer to improve derivatization efficiency.
  • Suitable for synthesis of ADC linkers and related bioconjugates.
  • Provided as a stable powder with defined cold-storage recommendations.
  • Offered in multiple pack sizes to support small-scale research and larger preparative work.
Catalog No. 50-002-03736