Learn More
Medchemexpress LLC DBCO-C6-acid | 1425485-72-8 | MFCD22380737 | 98.9% | 333.38 | C21H19NO3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12180550MG
Encompass_Preferred
DBCO-C6-acid is a dibenzocyclooctyne (DBCO)-containing click-chemistry reagent used as a non-cleavable linker in antibody-drug conjugate (ADC) synthesis and other bioconjugation workflows. Its strained alkyne enables copper-free, strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-functionalized partners, allowing efficient payload conjugation under mild conditions. The compound is supplied at high purity and has defined storage recommendations to support reproducible laboratory use.
  • Enables copper-free SPAAC conjugation with azide-functionalized partners.
  • Suitable for ADC linker synthesis and payload attachment workflows.
  • High reported purity for consistent and reproducible reactions.
  • Compatible with common organic solvents used in bioconjugation.
  • Stable under recommended storage conditions for reliable handling.
Catalog No. 50-002-02769