Learn More
Medchemexpress LLC DBCO-C6-acid | 1425485-72-8 | MFCD22380737 | 98.9% | 333.38 g/mol | C21H19NO3 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1218055G
Encompass_Preferred
DBCO-C6-acid is a dibenzocyclooctyne-containing reagent for copper-free click chemistry and bioconjugation. It functions as a non-cleavable linker or building block for synthesizing conjugates and for attaching payloads to azide-functionalized molecules via strain-promoted alkyne-azide cycloaddition (SPAAC). The reagent is provided as a solid with defined molecular weight and high purity to support reproducible conjugation workflows.
  • Enables copper-free SPAAC reactions with azide-functionalized partners.
  • Provides a six-carbon spacer for improved linker flexibility and conjugation efficiency.
  • High purity supports reproducible bioconjugation results.
  • Compatible with common organic solvents and standard conjugation protocols.
  • Suitable for synthesis of non-cleavable linkers and payload-linker constructs.
  • Available in small research-scale pack sizes for experimental workflows.
Catalog No. 50-002-01878