Learn More
Medchemexpress LLC DBCO-Cy3 | 1782950-79-1 | MFCD28505574 | 95.1% | 983.18 | C50H54N4O11S3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYD106910MG
Encompass_Preferred
DBCO-Cy3 is a DBCO-functionalized Cy3 fluorescent dye designed for copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) labeling of azide-containing biomolecules. It couples a dibenzocyclooctyne reactive group to an orange Cy3 fluorophore (excitation ~555 nm, emission ~580 nm), and is supplied with supporting documentation for analytical and safety data.
  • Enables copper-free SPAAC labeling of azide-containing biomolecules.
  • Orange fluorescence with excitation ~555 nm and emission ~580 nm.
  • Supplied as a 10 MG dry powder, soluble in DMSO.
  • High purity suitable for imaging applications.
  • Includes datasheet, COA, and SDS for quality and safety information.
Catalog No. 50-002-97701