Learn More
Medchemexpress LLC DBCO Ir catalyst | 2695594-15-9 | 93.1% | 1367.23 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1605235MG
Encompass_Preferred
DBCO Ir catalyst is an iridium catalyst labeled with DBCO. It can be used for in situ labeling and imaging of biomolecules (e.g., proteins, nucleic acids) in living organisms or for constructing targeted drug delivery systems. It acts as a catalyst in bioorthogonal reactions (e.g., copper-free click chemistry), facilitating the cycloaddition between DBCO and azide groups for specific labeling.
  • Iridium catalyst labeled with DBCO
  • Used for in situ labeling and imaging of biomolecules
  • Suitable for constructing targeted drug delivery systems
  • Acts as a catalyst in bioorthogonal reactions
  • Facilitates cycloaddition between DBCO and azide groups
Catalog No. 50-003-91452