Learn More
Medchemexpress LLC DBCO-Maleimide 5mg | 1395786-30-7 | 427.45 g/mol | C25H21N3O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1162705MG
Encompass_Preferred
DBCO-maleimide is a DBCO-functionalized maleimide linker used for bioconjugation and antibody-drug conjugate synthesis. It facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-bearing molecules, providing a stable, non-cleavable linkage. Supplied as a white to light yellow solid with high reported purity.
  • High purity (99.83%) suitable for sensitive conjugation applications.
  • Enables SPAAC click chemistry with azide-containing partners.
  • Provides a non-cleavable maleimide-DBCO linkage for stable conjugates.
  • Soluble in DMSO (≥ 100 mg/mL) for stock preparation.
  • Stored sealed at -20°C; in solution, keep at -80°C for long-term stability.
Catalog No. 50-002-29491