Learn More
Medchemexpress LLC DBCO-mPEG (MW 5kDa) | 99.10% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14031950MG
Encompass_Preferred
DBCO-mPEG (MW 5kDa) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is a click chemistry reagent, containing a DBCO group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains a DBCO group
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing azide groups
Catalog No. 50-003-57314