Learn More
Medchemexpress LLC DBCO-N-bis(PEG4-NHS ester) | 2639395-38-1 | 96.2% | 995.03 g/mol | C49H62N4O18 | 10 MG

Supplier: Medchemexpress LLC HY14509010MG
DBCO-N-bis(PEG4-NHS ester) is a PEG-based bifunctional linker that contains a dibenzocyclooctyne (DBCO) group and two PEG4-NHS ester reactive ends. It is designed for click chemistry applications, enabling strain-promoted alkyne-azide cycloaddition (SPAAC) for protein modification, labeling, and use as a linker in conjugate syntheses.
- Contains two PEG4-NHS ester groups and one DBCO group.
- Enables strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
- Suitable for protein modification and labeling applications.
- Molecular weight 995.03 g/mol; formula C49H62N4O18.
- Purity approximately 96.2%.
- Available in small quantities (for example, 10 mg) for laboratory use.
- Storage: pure form -20 °C (long-term); in solvent -80 °C (recommended for extended storage).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.