Learn More
Medchemexpress LLC DBCO-NHCO-PEG2-maleimide | 2698339-31-8 | 97.7% | C32H34N4O7 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13337210MG
Encompass_Preferred
DBCO-NHCO-PEG2-maleimide is a bifunctional click-chemistry linker that couples a dibenzocyclooctyne (DBCO) moiety for strain-promoted alkyne-azide cycloaddition (SPAAC) with a maleimide group for thiol-selective conjugation. The PEG2 spacer improves solubility and flexibility, making it suitable for bioconjugation and linker synthesis.
  • Enables copper-free SPAAC click chemistry
  • Provides thiol-selective maleimide conjugation
  • PEG2 spacer improves aqueous solubility
  • Suitable for bioconjugation and linker synthesis
  • High reported purity (~97.7%)
  • Stable as powder at -20°C for long-term storage
Catalog No. 50-002-59480