Learn More
Medchemexpress LLC Dbco-NHCO-S-S-NHS ester | 96.0% | 565.66 | C28H27N3O6S2 | 10 MG

Supplier: Medchemexpress LLC HY13341210MG
DBCO-NHCO-S-S-NHS ester is a cleavable linker used for assembling antibody-drug conjugates and for bioconjugation via strain-promoted alkyne-azide cycloaddition (SPAAC). It provides an NHS ester for amine coupling and a disulfide cleavable spacer for payload release.
- Molecular weight 565.66.
- Chemical formula C28H27N3O6S2.
- Purity approximately 96.0%.
- Physical form: solid, white to off-white.
- Contains a DBCO moiety for SPAAC click chemistry.
- Features an activated NHS ester for amine conjugation.
- Disulfide spacer provides cleavable linkage for payload release.
- Storage: powder at -20°C; in solvent at -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.