Learn More
Cayman Chemical DBCO-NHS ester 100mg
SDP
Supplier:  Cayman Chemical 40461100 MG
Encompass_Preferred
An azido-reactive click-chemistry reagent; has been conjugated to various anticancer agents for targeting azido-labeled cancer cells in mice
Catalog No. 50-294-4144