Learn More
Medchemexpress LLC DBCO-NHS ester 2 10mg | 1384870-47-6 | 430.45 g/mol | C25H22N2O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11552410MG
Encompass_Preferred
DBCO-NHS ester 2 is an amine-reactive dibenzocyclooctyne (DBCO) N-hydroxysuccinimide ester designed for efficient bioconjugation and copper-free click chemistry. It is provided as a small, research-scale solid with high reported purity and characterized molecular properties.
  • Amine-reactive NHS ester for rapid conjugation to primary amines.
  • DBCO handle enables copper-free strain-promoted azide-alkyne cycloaddition (SPAAC).
  • Suitable for antibody, protein, and small-molecule labeling workflows.
  • High purity supports reproducible conjugation and analytical characterization.
  • Supplied as a compact solid quantity for research-scale experiments.
Catalog No. 50-002-27925