Learn More
Medchemexpress LLC DBCO-NHS ester 3 25mg | 1393350-27-0 | 416.43 g·mol⁻1 | C24H20N2O5 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11554525MG
Encompass_Preferred
DBCO-NHS ester 3 is a DBCO-derived N-hydroxysuccinimidyl ester used as a click-chemistry reagent and ADC linker for bioconjugation. It combines an NHS ester for amine coupling with a DBCO moiety for strain-promoted azide-alkyne cycloaddition, enabling efficient labeling and antibody-drug conjugate synthesis.
  • Reacts with primary amines via the NHS ester for stable amide bond formation.
  • Enables strain-promoted azide-alkyne cycloaddition (SPAAC) through the DBCO moiety.
  • Intended for bioconjugation and antibody-drug conjugate linker synthesis.
  • Reported purity 95.55%.
  • Solid, white to off-white; powder storage -20°C up to 3 years.
Catalog No. 50-002-28806