Learn More
TARGETMOL CHEMICALS INC DBCO-NHS ester 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1506750MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. DBCO-NHS ester is an ADC linker, cleavable 3-unit PEG, which can also be used to synthesize antibody drug conjugates (ADCs). Purity 98.02%
Catalog No. 50-288-0036