Learn More
Medchemexpress LLC DBCO-(PEG)3-VC-PAB-MMAE | 2754384-60-4 | 99.6% | 1613.97 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY1110121MG 5
Encompass_Preferred
DBCO-(PEG)3-VC-PAB-MMAE is an agent-linker conjugate for ADC, formed by conjugating Monomethyl auristatin E to a DBCO-(PEG)3-vc-PAB linker. It is intended for cancer research and functions as a click chemistry reagent, utilizing a DBCO group for strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.
  • Agent-linker conjugate for ADC
  • Made by Monomethyl auristatin E conjugated to a DBCO-(PEG)3-vc-PAB linker
  • Click chemistry reagent
  • Contains a DBCO group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups
  • Can be used for the research of cancer
Catalog No. 50-003-43245